CAS RN 19879-30-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bavachinin, MedChemExpress
MedChemExpress Bavachinin(7-O-Methylbavachin) is a natural compound isolated from the Chinese herb Fructus Psoraleae;has potent anti-angiogenic activity.