accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 198649-68-2

FFFOBr

CAS RN 198649-68-2

IUPAC Name: 1-(bromomethyl)-2-(trifluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: TUNSVUOTVLWNQT-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results