accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 198560-43-9

OHO(S)FFNHOO

CAS RN 198560-43-9

IUPAC Name: (2S)-3-(3,4-difluorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-(3,4-difluorophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 423.42
Molecular Formula: C24H19F2NO4
InChi Key: IHSYIDJNVXPQRM-QFIPXVFZSA-N
SMILES: OC(=O)[C@H](CC1=CC=C(F)C(F)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results