accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 198419-91-9

CH3OOHOOHNH2

CAS RN 198419-91-9

IUPAC Name: 4-[amino(carboxy)methyl]-3-methylbenzoic acid
Synonyms: 4-[amino(carboxy)methyl]-3-methylbenzoic acid
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: SGIKDIUCJAUSRD-UHFFFAOYNA-N
SMILES: CC1=C(C=CC(=C1)C(O)=O)C(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results