accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19838-08-5

IUPAC Name: 3-methylpyrazin-2-amine
Synonyms: 3-methylpyrazin-2-amine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: VQPHZDDLWFHRHR-UHFFFAOYSA-N
SMILES: CC1=NC=CN=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results