accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

PSClClCl

CAS RN 1983-27-3

IUPAC Name: (chloromethyl)phosphonothioyl dichloride
Synonyms: chloromethylphosphonothioyl dichloride
Molecular Weight (g/mol): 183.41
Molecular Formula: CH2Cl3PS
InChi Key: HRHKZXIMFUYRDS-UHFFFAOYSA-N
SMILES: ClCP(Cl)(Cl)=S
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results