accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 198078-57-8

SNHO

CAS RN 198078-57-8

IUPAC Name: [6-(thiophen-2-yl)pyridin-3-yl]methanol
Synonyms: [6-(thiophen-2-yl)pyridin-3-yl]methanol
Molecular Weight (g/mol): 191.25
Molecular Formula: C10H9NOS
InChi Key: OLHQRPWNQRBQMA-UHFFFAOYSA-N
SMILES: OCC1=CN=C(C=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results