accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOHOO

CAS RN 19806-17-8

IUPAC Name: 2-[3-(carboxymethyl)phenyl]acetic acid
Synonyms: [3-(carboxymethyl)phenyl]acetic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: GDYYIJNDPMFMTB-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(CC(O)=O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)