accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19805-75-5

IUPAC Name: 3-carbamoyl-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrol-1-ium
Synonyms: 3-carbamoyl-2,2,5,5-tetramethyl-1H-pyrrol-1-ium
Molecular Weight (g/mol): 169.25
Molecular Formula: C9H17N2O
InChi Key: ACFYUJLIWIDSFM-UHFFFAOYSA-O
SMILES: CC1(C)[NH2+]C(C)(C)C(=C1)C(N)=O
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results