accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 197245-19-5

OOHNHOOSOO

CAS RN 197245-19-5

IUPAC Name: 2-({[(1,1-dioxo-1λ⁶-benzothiophen-2-yl)methoxy]carbonyl}amino)-3-phenylpropanoic acid
Synonyms: 2-({[(1,1-dioxo-1λ⁶-benzothiophen-2-yl)methoxy]carbonyl}amino)-3-phenylpropanoic acid
Molecular Weight (g/mol): 387.41
Molecular Formula: C19H17NO6S
InChi Key: DWLKZXPISLVRER-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC2=CC=CC=C2S1(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results