accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClOCl(E)

CAS RN 19672-59-4

IUPAC Name: (2E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one
Synonyms: (2E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one
Molecular Weight (g/mol): 277.14
Molecular Formula: C15H10Cl2O
InChi Key: YMEMCRBNZSLQCQ-XCVCLJGOSA-N
SMILES: ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results