CAS RN 195733-43-8
CAS RN 195733-43-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Atrasentan hydrochloride, MedChemExpress
MedChemExpress Atrasentan hydrochloride (ABT-627 hydrochloride) is a selective endothelin A receptor antagonist with an IC50 of 0.0551 nM for ETA.