accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19549-71-4

H3CH3COHCH3H3C

CAS RN 19549-71-4

IUPAC Name: 2,3-dimethylheptan-3-ol
Synonyms: 2,3-dimethylheptan-3-ol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: JIEGVNXCNNWVPH-UHFFFAOYNA-N
SMILES: CCCCC(C)(O)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results