accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OONHNH3

CAS RN 195314-59-1

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}cyclohexan-1-aminium
Synonyms: 4-[(tert-butoxycarbonyl)amino]cyclohexan-1-aminium
Molecular Weight (g/mol): 215.32
Molecular Formula: C11H23N2O2
InChi Key: FEYLUKDSKVSMSZ-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)NC1CCC([NH3+])CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results