accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NClHCl

CAS RN 19524-08-4

IUPAC Name: 4-chloro-3-methylpyridine hydrochloride
Synonyms: 4-chloro-3-methylpyridine hydrochloride
Molecular Weight (g/mol): 164.03
Molecular Formula: C6H7Cl2N
InChi Key: WPIUSHWGBZMBHZ-UHFFFAOYSA-N
SMILES: Cl.CC1=C(Cl)C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results