accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1949-78-6

OHOOHHOHO

CAS RN 1949-78-6

IUPAC Name: oxane-2,3,4,5-tetrol
Synonyms: xylose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: SRBFZHDQGSBBOR-UHFFFAOYNA-N
SMILES: OC1COC(O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results