accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 194240-75-0

OHOClF

CAS RN 194240-75-0

IUPAC Name: 2-(4-chloro-2-fluorophenyl)acetic acid
Synonyms: (4-chloro-2-fluorophenyl)acetic acid
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: WKAZXGAJEGPUAY-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(Cl)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results