accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19423-87-1

ClClClYb

CAS RN 19423-87-1

IUPAC Name: ytterbium(3+) hydrate trichloride
Synonyms: ytterbium(3+) hydrate trichloride
Molecular Weight (g/mol): 297.42
Molecular Formula: Cl3H2OYb
InChi Key: WGKCQSFQBYKUTC-UHFFFAOYSA-K
SMILES: O.[Cl-].[Cl-].[Cl-].[Yb+3]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)