CAS RN 19423-85-9
CAS RN 19423-85-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Erbium(III) chloride hydrate, REacton™, 99.9% (REO)
CAS: 19423-85-9 Molecular Formula: Cl3Er Molecular Weight (g/mol): 273.61 MDL Number: MFCD00010989 InChI Key: HDGGAKOVUDZYES-UHFFFAOYSA-K IUPAC Name: erbium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Er+3]
2
Erbium(III) chloride hydrate, REacton™, 99.99% (REO)
CAS: 19423-85-9 Molecular Formula: Cl3Er Molecular Weight (g/mol): 273.61 MDL Number: MFCD00010989 InChI Key: HDGGAKOVUDZYES-UHFFFAOYSA-K IUPAC Name: erbium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Er+3]