accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOONOO

CAS RN 193693-64-0

IUPAC Name: 1-{[(9H-fluoren-9-yl)methoxy]carbonyl}azetidine-3-carboxylic acid
Synonyms: 1-[(9H-fluoren-9-ylmethoxy)carbonyl]azetidine-3-carboxylic acid
Molecular Weight (g/mol): 323.35
Molecular Formula: C19H17NO4
InChi Key: QDEJCUHGSHSYQH-UHFFFAOYSA-N
SMILES: OC(=O)C1CN(C1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)