accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOO(S)NOO

CAS RN 193693-60-6

IUPAC Name: 2-[(2S)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidin-2-yl]acetic acid
Synonyms: [(2S)-1-[(9H-fluoren-9-ylmethoxy)carbonyl]pyrrolidin-2-yl]acetic acid
Molecular Weight (g/mol): 351.4
Molecular Formula: C21H21NO4
InChi Key: ZNIGOUDZWCDFFC-AWEZNQCLSA-N
SMILES: OC(=O)C[C@@H]1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results