CAS RN 193551-21-2
CAS RN 193551-21-2
IUPAC Name:
4-[4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl]cyclohexan-1-ol
Synonyms:
4-[4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)imidazol-1-yl]cyclohexan-1-ol
Molecular Weight (g/mol):
368.41
Molecular Formula:
C20H21FN4O2
InChi Key:
ZQUSFAUAYSEREK-UHFFFAOYSA-N
SMILES:
COC1=NC=CC(=N1)C1=C(N=CN1C1CCC(O)CC1)C1=CC=C(F)C=C1
1
–
2
of
2
results
2
SB 239063, MedChemExpress
MedChemExpress SB 239063 is a potent, selective and orally active p38 MAPK inhibitor, exhibits an IC50 of 44 nM for recombinant purified human p38α, with equipotent inhibitory activity against p38α and p38β. SB 239063 has no effect on p38γ or p38δ. With anti-asthma activity and also be used to enhance memory which is impaired due to aging or medical conditions, such as, AD.