accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2HNN

CAS RN 19335-11-6

IUPAC Name: 1H-indazol-5-amine
Synonyms: 1H-indazol-5-amine
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: XBTOSRUBOXQWBO-UHFFFAOYSA-N
SMILES: NC1=CC=C2NN=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results