accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19315-93-6

HOHNO

CAS RN 19315-93-6

IUPAC Name: 6-hydroxy-1,2-dihydroquinolin-2-one
Synonyms: 6-hydroxy-2-quinolone
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: AQLYZDRHNHZHIS-UHFFFAOYSA-N
SMILES: OC1=CC=C2NC(=O)C=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results