accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2SNHClCl

CAS RN 19250-09-0

IUPAC Name: (3,4-dichlorophenyl)thiourea
Synonyms: 3,4-dichlorophenylthiourea
Molecular Weight (g/mol): 221.1
Molecular Formula: C7H6Cl2N2S
InChi Key: CCNCITSJXCSXJY-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results