accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19230-28-5

ClClI

CAS RN 19230-28-5

IUPAC Name: 1,3-dichloro-2-iodobenzene
Synonyms: 1,3-dichloro-2-iodobenzene
Molecular Weight (g/mol): 272.89
Molecular Formula: C6H3Cl2I
InChi Key: ZMPGXSFTXBOKFM-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1I
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results