accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19228-91-2

H3CH3CCH3OONHS

CAS RN 19228-91-2

IUPAC Name: tert-butyl N-(thiophen-3-yl)carbamate
Synonyms: tert-butyl N-(thiophen-3-yl)carbamate
Molecular Weight (g/mol): 199.27
Molecular Formula: C9H13NO2S
InChi Key: PRWYQCYSADTIBZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results