accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NHOOFPFFFFF

CAS RN 19200-21-6

IUPAC Name: dioxoamine; hexafluoro-λ⁵-phosphanuide
Synonyms: nitrite ion hexafluorophosphate
Molecular Weight (g/mol): 191.98
Molecular Formula: F6HNO2P
InChi Key: WCPUJULSPZCGOX-UHFFFAOYSA-N
SMILES: [O-][NH+]=O.F[P-](F)(F)(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results