CAS RN 191671-46-2
CAS RN 191671-46-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Sulfo-NHS-LC-Biotin
Amine reactive biotinylation reagent, cell impermeable