accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 191604-88-3

H3COOFFFOHO

CAS RN 191604-88-3

IUPAC Name: 2-methoxy-5-(trifluoromethoxy)benzoic acid
Synonyms: 2-methoxy-5-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 236.15
Molecular Formula: C9H7F3O4
InChi Key: HURBWIHJHDFCGU-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC(F)(F)F)C=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results