accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONOONCS

CAS RN 190774-55-1

IUPAC Name: 1-isothiocyanato-2-methoxy-4-nitrobenzene
Synonyms: 1-isothiocyanato-2-methoxy-4-nitrobenzene
Molecular Weight (g/mol): 210.21
Molecular Formula: C8H6N2O3S
InChi Key: NXWXXLFRMVILJN-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(=C1)[N+]([O-])=O)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results