accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19056-40-7

H3COH2NBr

CAS RN 19056-40-7

IUPAC Name: 4-bromo-3-methoxyaniline
Synonyms: 4-bromo-3-methoxyaniline
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: RUTNWXBHRAIQSP-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=CC(N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results