accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2HNOH2N

CAS RN 1904-58-1

IUPAC Name: 2-aminobenzohydrazide
Synonyms: benzoylhydrazine, O-amino
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: GRWMSCBKWMQPON-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results