accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 189807-20-3

FFClOF

CAS RN 189807-20-3

IUPAC Name: 2,3,6-trifluorobenzoyl chloride
Synonyms: 2,3,6-trifluorobenzoyl chloride
Molecular Weight (g/mol): 194.54
Molecular Formula: C7H2ClF3O
InChi Key: DMUGJQMADYEWSQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(C(Cl)=O)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results