CAS RN 189744-94-3
CAS RN 189744-94-3
IUPAC Name:
2-methyl-5-[(4-phenyl-1,2,3,6-tetrahydropyridin-1-yl)methyl]pyrimidin-4-amine dihydrochloride
Synonyms:
2-methyl-5-[(4-phenyl-3,6-dihydro-2H-pyridin-1-yl)methyl]pyrimidin-4-amine dihydrochloride
Molecular Weight (g/mol):
353.29
Molecular Formula:
C17H22Cl2N4
InChi Key:
UZHMRJRDYCRKIZ-UHFFFAOYSA-N
SMILES:
Cl.Cl.CC1=NC=C(CN2CCC(=CC2)C2=CC=CC=C2)C(N)=N1
1
–
1
of
1
results
1
Ro 10-5824 dihydrochloride, MedChemExpress
MedChemExpress Ro 10-5824 dihydrochloride is a selective dopamine D4 receptor partial agonist, with Ki of 5.2 nM.