accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1889-78-7

OClBr

CAS RN 1889-78-7

IUPAC Name: 2-bromo-1-(4-chlorophenyl)-2-phenylethan-1-one
Synonyms: 2-bromo-1-(4-chlorophenyl)-2-phenylethanone
Molecular Weight (g/mol): 309.59
Molecular Formula: C14H10BrClO
InChi Key: OBEFSOTWERFWSZ-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C(Br)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results