accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OMoO

CAS RN 18868-43-4

IUPAC Name: dioxomolybdenum
Synonyms: molybdenum(IV) oxide
Molecular Weight (g/mol): 127.95
Molecular Formula: MoO2
InChi Key: QXYJCZRRLLQGCR-UHFFFAOYSA-N
SMILES: O=[Mo]=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results