accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 188589-61-9

CH3ONHFFFONH3CCH3

CAS RN 188589-61-9

IUPAC Name: 1,4a,6a-trimethyl-2-oxo-N-[2-(trifluoromethyl)phenyl]-1H,2H,4aH,4bH,5H,6H,6aH,7H,8H,9H,9aH,9bH,10H,11H,11aH-indeno[5,4-f]quinoline-7-carboxamide
Synonyms: 1,4a,6a-trimethyl-2-oxo-N-[2-(trifluoromethyl)phenyl]-4bH,5H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-indeno[5,4-f]quinoline-7-carboxamide
Molecular Weight (g/mol): 474.57
Molecular Formula: C27H33F3N2O2
InChi Key: YFBLEKKYWFJKBP-UHFFFAOYNA-N
SMILES: CN1C2CCC3C4CCC(C(=O)NC5=CC=CC=C5C(F)(F)F)C4(C)CCC3C2(C)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results