accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 18858-05-4

CH3ClSH

CAS RN 18858-05-4

IUPAC Name: 2-chloro-6-methylbenzene-1-thiol
Synonyms: 2-chloro-6-methylbenzenethiol
Molecular Weight (g/mol): 158.64
Molecular Formula: C7H7ClS
InChi Key: WNRLJMYSWRBJIG-UHFFFAOYSA-N
SMILES: CC1=C(S)C(Cl)=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)