accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3H3COONHCH3HOOOH3C

CAS RN 188476-33-7

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxy-2-methylpropanoate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]-3-hydroxy-2-methylpropanoate
Molecular Weight (g/mol): 233.26
Molecular Formula: C10H19NO5
InChi Key: OUUNEDPIBZNRMT-UHFFFAOYNA-N
SMILES: COC(=O)C(C)(CO)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results