accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 18800-28-7

BrOBr

CAS RN 18800-28-7

IUPAC Name: 1-bromo-2-(2-bromoethoxy)benzene
Synonyms: 1-bromo-2-(2-bromoethoxy)benzene
Molecular Weight (g/mol): 279.96
Molecular Formula: C8H8Br2O
InChi Key: LOVVQTBEUIBASP-UHFFFAOYSA-N
SMILES: BrCCOC1=CC=CC=C1Br
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)