accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNClOOH

CAS RN 187998-35-2

IUPAC Name: 1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
Synonyms: 1-(4-chlorophenyl)-5-methylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 236.66
Molecular Formula: C11H9ClN2O2
InChi Key: OAPQKGGVDCZENK-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1C1=CC=C(Cl)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results