accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1878-85-9

CH3OOHOO

CAS RN 1878-85-9

IUPAC Name: 2-(2-methoxyphenoxy)acetic acid
Synonyms: O-methoxy phenoxyacetic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: IHONYPFTXGQWAX-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)