accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1869-22-3

FFFHNNH2Cl

CAS RN 1869-22-3

IUPAC Name: [2-chloro-5-(trifluoromethyl)phenyl]hydrazine
Synonyms: [2-chloro-5-(trifluoromethyl)phenyl]hydrazine
Molecular Weight (g/mol): 210.58
Molecular Formula: C7H6ClF3N2
InChi Key: PAPSYPXQNRTIRR-UHFFFAOYSA-N
SMILES: NNC1=C(Cl)C=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results