accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 18686-82-3

IUPAC Name: 2,3-dihydro-1,3,4-thiadiazole-2-thione
Synonyms: 3H-1,3,4-thiadiazole-2-thione
Molecular Weight (g/mol): 118.17
Molecular Formula: C2H2N2S2
InChi Key: JLAMDELLBBZOOX-UHFFFAOYSA-N
SMILES: S=C1NN=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results