accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1868-58-2

ONONFBr

CAS RN 1868-58-2

IUPAC Name: 3-bromo-2-fluoro-5-nitropyridine
Synonyms: 3-bromo-2-fluoro-5-nitropyridine
Molecular Weight (g/mol): 220.99
Molecular Formula: C5H2BrFN2O2
InChi Key: NLLPMLSUMJHTCJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(F)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results