accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FClClOFFF

CAS RN 186517-45-3

IUPAC Name: 3-chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride
Synonyms: 3-chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 261
Molecular Formula: C8H2Cl2F4O
InChi Key: QRJJXSXDJDIRPI-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC(=C1C(Cl)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results