accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1864-94-4

OO

CAS RN 1864-94-4

IUPAC Name: phenyl formate
Synonyms: anisic aldehyde
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: GEOWCLRLLWTHDN-UHFFFAOYSA-N
SMILES: O=COC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results