accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNOOH

CAS RN 186129-25-9

IUPAC Name: 1-methyl-1H-indole-5-carboxylic acid
Synonyms: 1-methylindole-5-carboxylic acid
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: UHQAIJFIXCOBCN-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC(=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)