accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1858240-14-8

NNClBrOHOH2O

CAS RN 1858240-14-8

IUPAC Name: 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Synonyms: 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Molecular Weight (g/mol): 293.5
Molecular Formula: C8H6BrClN2O3
InChi Key: RYTIKZUUZLHHDR-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=C(Br)N2C=C(Cl)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results